missing translation for 'onlineSavingsMsg'
Learn More

Homovanillic Acid-13C6, TRC

Product Code. 30393075
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
10 mg
Unit Size:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393075 10 mg -
30401908 0.5 mg 0.5mg
30406612 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393075 Supplier Toronto Research Chemicals Supplier No. TRCH66950310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185016-45-8
Molecular Formula 13C6 C3 H10 O4
Molecular Weight (g/mol) 188.1292
Synonym Benzene-1,2,3,4,5,6-13C6-acetic acid, 4-hydroxy-3-methoxy-, 2-(4-Hydroxy-3-methoxyphenyl-1,2,3,4,5,6-13C6)acetic acid, 4-Hydroxy-3-methoxybenzene-1,2,3,4,5,6-13C6-acetic acid, (4-Hydroxy-3-methoxyphenyl-13C6)acetic acid, [3-Methoxy-4-hydroxy(phenyl-13C6)]acetic acid, 3'-Methoxy-4'-hydroxyphenyl-13C6-acetic acid, Homovanillic acid-13C6, Homovanillic acid 13C6 (phenyl-13C6), Vanilacetic acid-13C6
IUPAC Name 2-(4-hydroxy-3-methoxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)acetic acid
SMILES CO[13c]1[13cH][13c](CC(=O)O)[13cH][13cH][13c]1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H10O4/c1-13-8-4-6(5-9(11)12)2-3-7(8)10/h2-4,10H,5H2,1H3,(H,11,12)/i2+1,3+1,4+1,6+1,7+1,8+1
Formula Weight 188.078
Percent Purity >95%
Chemical Name or Material Homovanillic acid-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.