CAS RN 1185016-45-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Homovanillic Acid-13C6, TRC
CAS: 1185016-45-8 Molecular Formula: 13C6 C3 H10 O4 Molecular Weight (g/mol): 188.1292 Synonym: Benzene-1,2,3,4,5,6-13C6-acetic acid, 4-hydroxy-3-methoxy-,2-(4-Hydroxy-3-methoxyphenyl-1,2,3,4,5,6-13C6)acetic acid,4-Hydroxy-3-methoxybenzene-1,2,3,4,5,6-13C6-acetic acid,(4-Hydroxy-3-methoxyphenyl-13C6)acetic acid,[3-Methoxy-4-hydroxy(phenyl-13C6)]acetic acid,3'-Methoxy-4'-hydroxyphenyl-13C6-acetic acid,Homovanillic acid-13C6,Homovanillic acid 13C6 (phenyl-13C6),Vanilacetic acid-13C6 IUPAC Name: 2-(4-hydroxy-3-methoxy(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-yl)acetic acid SMILES: CO[13c]1[13cH][13c](CC(=O)O)[13cH][13cH][13c]1O