missing translation for 'onlineSavingsMsg'
Learn More

Homoharringtonine, TRC

Product Code. 30403049
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403049 50 mg
30407657 25 mg
30389890 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403049 Supplier Toronto Research Chemicals Supplier No. TRCH59650050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26833-87-4
Molecular Formula C29 H39 N O9
Molecular Weight (g/mol) 545.62
Synonym Cephalotaxine, 3-[4-methyl (2R)-2-hydroxy-2-(4-hydroxy-4-methylpentyl)butanedioate], Cephalotaxine, 4-methyl (2R)-2-hydroxy-2-(4-hydroxy-4-methylpentyl)butanedioate (ester) (9CI), Homoharringtonine (8CI), CGX 635, Ceflatonin, Myelostat, NSC 141633, Omacetaxine mepesuccinate, Synribo
SMILES COC(=O)C[C@](O)(CCCC(C)(C)O)C(=O)O[C@H]1[C@H]2c3cc4OCOc4cc3CCN5CCC[C@]25C=C1OC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H39NO9/c1-27(2,33)8-5-10-29(34,16-23(31)36-4)26(32)39-25-22(35-3)15-28-9-6-11-30(28)12-7-18-13-20-21(38-17-37-20)14-19(18)24(25)28/h13-15,24-25,33-34H,5-12,16-17H2,1-4H3/t24-,25-,28+,29-/m1/s1
Formula Weight 545.2625
Chemical Name or Material Homoharringtonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.