accessibility menu, dialog, popup

UserName

CH3OOHOH3CH3COHOO(S)(R)OON(R)OH3C

CAS RN 26833-87-4

IUPAC Name: (2R,3S,6R)-4-methoxy-16,18-dioxa-10-azapentacyclo[11.7.0.0²,⁶.0⁶,¹⁰.0¹⁵,¹⁹]icosa-1(20),4,13,15(19)-tetraen-3-yl 1-methyl 3-hydroxy-3-(4-hydroxy-4-methylpentyl)butanedioate
Synonyms: omacetaxine mepesuccinate
Molecular Weight (g/mol): 545.63
Molecular Formula: C29H39NO9
InChi Key: HYFHYPWGAURHIV-ZEDNPHJLSA-N
SMILES: COC(=O)CC(O)(CCCC(C)(C)O)C(=O)O[C@H]1[C@@H]2C3=CC4=C(OCO4)C=C3CCN3CCC[C@]23C=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)