missing translation for 'onlineSavingsMsg'
Learn More

Homo Sildenafil-d5, TRC

Product Code. 30392059
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392059 1 mg
30403048 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392059 Supplier Toronto Research Chemicals Supplier No. TRCH6151521MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216711-61-3
Molecular Formula C23H27D5N6O4S
Molecular Weight (g/mol) 493.63
Synonym 5-[2-Ethoxy-5-[(4-(ethyl-d5)-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one
IUPAC Name 5-(2-ethoxy-5-((4-(ethyl-d5)piperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES O=C(N1)C2=C(C(CCC)=NN2C)N=C1C3=CC(S(N4CCN(C([2H])([2H])C([2H])([2H])[2H])CC4)(=O)=O)=CC=C3OCC

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C23H32N6O4S/c1-5-8-18-20-21(27(4)26-18)23(30)25-22(24-20)17-15-16(9-10-19(17)33-7-3)34(31,32)29-13-11-28(6-2)12-14-29/h9-10,15H,5-8,11-14H2,1-4H3,(H,24,25,30)/i2D3,6D2
Formula Weight 493.24
Chemical Name or Material Homo Sildenafil-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.