CAS RN 1216711-61-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Homo Sildenafil-d5, TRC
CAS: 1216711-61-3 Molecular Formula: C23H27D5N6O4S Molecular Weight (g/mol): 493.63 Synonym: 5-[2-Ethoxy-5-[(4-(ethyl-d5)-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one IUPAC Name: 5-(2-ethoxy-5-((4-(ethyl-d5)piperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one SMILES: O=C(N1)C2=C(C(CCC)=NN2C)N=C1C3=CC(S(N4CCN(C([2H])([2H])C([2H])([2H])[2H])CC4)(=O)=O)=CC=C3OCC