missing translation for 'onlineSavingsMsg'
Learn More

Homo-L-tyrosine Hydrobromide, TRC

Product Code. 30399466
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399466 250 mg 250mg
30398790 100 mg 100mg
30402941 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399466 Supplier Toronto Research Chemicals Supplier No. TRCH668000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141899-12-9
Molecular Formula C10H14BrNO3
Molecular Weight (g/mol) 276.13
Synonym (alphaS)-Amino-4-hydroxybenzenebutanoic Acid Hydrobromide
IUPAC Name (2S)-2-amino-4-(4-hydroxyphenyl)butanoic acid;hydrobromide
SMILES Br.N[C@@H](CCc1ccc(O)cc1)C(=O)O

Specifications

InChI Formula InChI=1S/C10H13NO3.BrH/c11-9(10(13)14)6-3-7-1-4-8(12)5-2-7;/h1-2,4-5,9,12H,3,6,11H2,(H,13,14);1H/t9-;/m0./s1
Quantity 250 mg
Formula Weight 275.0157
Shipping Condition Room Temperature
Chemical Name or Material Homo-L-tyrosine Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.