CAS RN 141899-12-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Homo-L-tyrosine Hydrobromide, TRC
CAS: 141899-12-9 Molecular Formula: C10H14BrNO3 Molecular Weight (g/mol): 276.13 Synonym: (alphaS)-Amino-4-hydroxybenzenebutanoic Acid Hydrobromide IUPAC Name: (2S)-2-amino-4-(4-hydroxyphenyl)butanoic acid;hydrobromide SMILES: Br.N[C@@H](CCc1ccc(O)cc1)C(=O)O