missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homez Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80267
This item is not returnable.
View return policy
Description
Homez Polyclonal specifically detects Homez in Mouse samples. It is validated for Western Blot.
Specifications
| Homez | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| homeobox and leucine zipper encoding, homeobox and leucine zipper protein Homez, KIAA1443Homeodomain leucine zipper-containing factor | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 57594 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_898997 | |
| HOMEZ | |
| Synthetic peptide directed towards the N terminal of mouse HOMEZ. Peptide sequence LSPLAPSEQPTHMKGLKVEPEEPSQVSQLPLNHQNAKEPLMMGSRTFSHQ. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Mouse: 100%; Rat: 84%. | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction