missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOIP/RNF31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | HOIP/RNF31 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18141893
|
Novus Biologicals
NBP2-27290SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18124754
|
Novus Biologicals
NBP2-27290 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HOIP/RNF31 Polyclonal specifically detects HOIP/RNF31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HOIP/RNF31 | |
| Western Blot, Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| 55072 | |
| A portion of amino acids 950-1000 from human RNF31 (HOIP) was used as the immunogen for this antibody. NP-060469.4 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein | |
| RNF31 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, 94% in new world monkey, horse, naked mole rat, crab-eating macaque, rhesus monkey, white-cheeked gibbon, common chimpanzee, and sumatran orangutan, 88% in guinea pig, dog, common rabbit, and pig, 82% in panda, Chinese hamster, african bush elephant, gray short-tailed opossum, platypus, rat, and clawed frog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title