missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOIP/RNF31 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27290
This item is not returnable.
View return policy
Description
HOIP/RNF31 Polyclonal specifically detects HOIP/RNF31 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HOIP/RNF31 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| RNF31 | |
| A portion of amino acids 950-1000 from human RNF31 (HOIP) was used as the immunogen for this antibody. NP-060469.4 | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, 94% in new world monkey, horse, naked mole rat, crab-eating macaque, rhesus monkey, white-cheeked gibbon, common chimpanzee, and sumatran orangutan, 88% in guinea pig, dog, common rabbit, and pig, 82% in panda, Chinese hamster, african bush elephant, gray short-tailed opossum, platypus, rat, and clawed frog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry-Paraffin 5ug/ml | |
| EC 6.3.2.-, FLJ10111, FLJ23501, HOIP, MGC19975, ring finger protein 31, ZIBRAHOIL-1-interacting protein, Zinc in-between-RING-finger ubiquitin-associated domain protein | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 55072 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction