missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for HE 99X Max Submarine Electrophoresis Unit

Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit

Brand:  Hoefer™ HE91A-30-1.0

Code : NEW

Additional Details : Weight : 0.09000kg

 View more versions of this product

Product Code. 10442972

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Requires : Comb back must be ordered separately (see ordering table)

Description

Description

Backless combs for use with Hoefer HE99X Max Submarine Electrophoresis Unit

Combs can be used interchangeably with a single comb back.

Specifications

Specifications

HE99X Max Submarine Electrophoresis Unit
30
0.1 cm
0.11 in.
0.19 in.
Backless Comb
0.03 in.
15 μL
0.3 cm
0.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers