missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hoechst 33258 trihydrochloride

Product Code. p-200104460
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16442717

Brand: MedChemExpress HY15558A50mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hoechst 33258 trihydrochloride is a fluorescent dyes, which can be used as a cell dye for DNA.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 50 mg
Shipping Condition Room temperature
Solubility H2O : ≥ 72.85 mg/mL (136.45 mM)
Molecular Weight (g/mol) 533.88 g/mol
CAS 23491-45-4
Synonym bisBenzimide H 33258 trihydrochloride ∣ H 33258 trihydrochloride
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.