missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hoechst 33258 trihydrochloride

Product Code. 16442717
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16442717 50 mg 50mg
16426806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16442717 Supplier MedChemExpress Supplier No. HY15558A50mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hoechst 33258 trihydrochloride is a fluorescent dyes, which can be used as a cell dye for DNA.

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Quantity 50 mg
Shipping Condition Room temperature
Solubility H2O : ≥ 72.85 mg/mL (136.45 mM)
Molecular Weight (g/mol) 533.88 g/mol
CAS 23491-45-4
Synonym bisBenzimide H 33258 trihydrochloride ∣ H 33258 trihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.