missing translation for 'onlineSavingsMsg'
Learn More

HNRPGT Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-33267-20ul

 View more versions of this product

Product Code. 30231670

  • £171.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HNRPGT Monoclonal antibody specifically detects HNRPGT in Human samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HNRPGT
Monoclonal
ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
hnRNP G-T, HNRNPG-T, HNRNPGTRNA-binding motif protein, X-linked-like-2, HNRPGTheterogeneous nuclear ribonucleoprotein G T, RNA binding motif protein, X-linked-like 2, testes specific heterogenous nuclear ribonucleoprotein G T, testes-specific heterogenous nuclear ribonucleoprotein G-T, Testis-specific heterogeneous nuclear ribonucleoprotein G-T
Recombinant fusion protein containing a sequence corresponding to amino acids 175-350 of human HNRPGT (NP_055284.3).,, Sequence:, GGMRGRALAVRGRDGYSGPPRREPLPPRRDPYLGPRDEGYSSRDGYSSRDYREPRGFAPSPGEYTHRDYGHSSVRDDCPLRGYSDRDGYGGRDRDYGDHLSRGSHREPFESYGELRGAAPGRGTPPSYGGGGRYEEYRGYSPDAYSGGRDSYSSSYGRSDRYSRGRHRVGRPDRGL
20 μL
Primary
Human
Purified
ELISA, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
27288
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.