missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRPGT Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£171.00 - £401.00
Specifications
| Antigen | HNRPGT |
|---|---|
| Dilution | ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231670
|
Novus Biologicals
NBP3-33267-20ul |
20 μL |
£171.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226774
|
Novus Biologicals
NBP3-33267-100ul |
100 μL |
£401.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HNRPGT Monoclonal antibody specifically detects HNRPGT in Human samples. It is validated for ELISA,Immunocytochemistry/ImmunofluorescenceSpecifications
| HNRPGT | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| hnRNP G-T, HNRNPG-T, HNRNPGTRNA-binding motif protein, X-linked-like-2, HNRPGTheterogeneous nuclear ribonucleoprotein G T, RNA binding motif protein, X-linked-like 2, testes specific heterogenous nuclear ribonucleoprotein G T, testes-specific heterogenous nuclear ribonucleoprotein G-T, Testis-specific heterogeneous nuclear ribonucleoprotein G-T | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 175-350 of human HNRPGT (NP_055284.3).,, Sequence:, GGMRGRALAVRGRDGYSGPPRREPLPPRRDPYLGPRDEGYSSRDGYSSRDYREPRGFAPSPGEYTHRDYGHSSVRDDCPLRGYSDRDGYGGRDRDYGDHLSRGSHREPFESYGELRGAAPGRGTPPSYGGGGRYEEYRGYSPDAYSGGRDSYSSSYGRSDRYSRGRHRVGRPDRGL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 27288 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title