missing translation for 'onlineSavingsMsg'
Learn More

HNRNPA1 Mouse, anti-Human, Unconjugated, Clone: 1A5A4, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen HNRNPA1
Clone 1A5A4
Concentration 500 μg/mL
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17271506
View Documents
Proteintech
67844-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17261506
View Documents
Proteintech
67844-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

In eukaryotic cells, nascent RNA polymerase II transcripts are associated in the nucleus with specific proteins to form ribonucleoprotein complexes called HNRP or 40S. Protein moiety of the HNRP particle has 6 major components called core proteins, and HNRPA1 is one of them. hnRNP A1 is a multifunctional RNA binding protein which is involved in pre-mRNA splicing, export of mature transcripts from the nucleus, mRNA turnover, and internal ribosome entry site (IRES)-mediated translation. HNRNPA1 has three isoforms with MW 30 kDa, 34 kDa and 39 kDa.

Specifications

Specifications

HNRNPA1
500 μg/mL
Monoclonal
Liquid
Human, Mouse
Primary
1A5A4
Western Blot, ELISA
Unconjugated
Mouse
3178
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.