Learn More
HNRNPA1 Mouse, anti-Human, Unconjugated, Clone: 1A5A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67844-1-IG-20UL
 View more versions of this product
Product Code. 17271506
  • £135.00 / 20µL
Description
In eukaryotic cells, nascent RNA polymerase II transcripts are associated in the nucleus with specific proteins to form ribonucleoprotein complexes called HNRP or 40S. Protein moiety of the HNRP particle has 6 major components called core proteins, and HNRPA1 is one of them. hnRNP A1 is a multifunctional RNA binding protein which is involved in pre-mRNA splicing, export of mature transcripts from the nucleus, mRNA turnover, and internal ribosome entry site (IRES)-mediated translation. HNRNPA1 has three isoforms with MW 30 kDa, 34 kDa and 39 kDa.
Specifications
Specifications
HNRNPA1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse
Western Blot, ELISA
1A5A4
Unconjugated
20 μL
3178