missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP M Antibody (2A6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
£171.00 - £370.00
Specifications
| Antigen | hnRNP M |
|---|---|
| Clone | 2A6 |
| Dilution | Western Blot 1:1000, Simple Western 1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18244880
|
Novus Biologicals
NB200-315SS |
0.025 mL |
£171.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18718243
|
Novus Biologicals
NB200-315 |
0.1 mL |
£370.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hnRNP M Monoclonal specifically detects hnRNP M in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| hnRNP M | |
| Western Blot 1:1000, Simple Western 1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine, Rabbit | |
| P52272 | |
| 4670 | |
| M19 fusion protein containing full-length human protein. [UniProt# P52272] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| 68 kDa |
| 2A6 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| Ascites with 0.1% Sodium Azide | |
| DKFZp547H118, heterogeneous nuclear ribonucleoprotein M, hnRNP M, HNRNPM4, HNRPM4heterogenous nuclear ribonucleoprotein M, HNRPMheterogenous nuclear ribonucleoprotein M4, HTGR1, M4 protein, N-acetylglucosamine receptor 1, NAGR1hnRNA-binding protein M4 | |
| HNRNPM | |
| IgG2b κ | |
| Unpurified | |
| NB200-315 is specific for hnRNP M3 and M4 proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title