missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP M Antibody (2A6), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB200-315SS
This item is not returnable.
View return policy
Description
hnRNP M Monoclonal specifically detects hnRNP M in Human, Mouse, Rat, Porcine, Bovine, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| hnRNP M | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| DKFZp547H118, heterogeneous nuclear ribonucleoprotein M, hnRNP M, HNRNPM4, HNRPM4heterogenous nuclear ribonucleoprotein M, HNRPMheterogenous nuclear ribonucleoprotein M4, HTGR1, M4 protein, N-acetylglucosamine receptor 1, NAGR1hnRNA-binding protein M4 | |
| Mouse | |
| 68 kDa | |
| 0.025 mL | |
| Cancer | |
| 4670 | |
| Human, Mouse, Rat, Pig, Bovine, Rabbit | |
| Ascites |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 2A6 | |
| Western Blot 1:1000, Simple Western 1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| P52272 | |
| HNRNPM | |
| M19 fusion protein containing full-length human protein. [UniProt# P52272] | |
| Unpurified | |
| RUO | |
| Primary | |
| NB200-315 is specific for hnRNP M3 and M4 proteins. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction