missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HNF6 Recombinant Rabbit Monoclonal Antibody (7L3O1)

Rabbit Recombinant Monoclonal Antibody

£147.00 - £363.00

Specifications

Antigen HNF6
Clone 7L3O1
Concentration 2.29 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30282706
View Documents
Invitrogen™
MA558123
20 μL
£147.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30282712
View Documents
Invitrogen™
MA558124
100 μL
£363.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunogen sequence: PYHKDVAGMG QSLSPLSSSG LGSIHNSQQG LPHYAHPGAA MPTDKMLTPN GFEAHHPAML GRHGEQHLTP TSAGMVPING LPPHHPHAHL NAQGHGQLLG TAREPNPSVT GAQVSNGSNS GQMEEI.

Hepatocyte nuclear factor 6 is a CUT homeobox family protein which is a transcription factor and contains a CUT DNA-binding domain and a homeobox DNA-binding domain. It is an upstream regulator of several genes involved in the pathogenesis of maturity-onset diabetes of the young. HNF6 is a ligand of the FOXM1 transcription factor network. HNF6, along with HNF4, controls gene expression in the liver. The expression levels of HNF6 and HNF4 rise steadily during development and that up- or down regulation of HNF6 and HNF4 expression generate premature or delayed expression of the g6pc gene.
Specifications

Specifications

HNF6
2.29 mg/mL
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
RUO
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
O08755, P70512
15379, 25231
Recombinant fusion protein containing a sequence corresponding to amino acids 167-292 of human HNF-6/ONECUT1 (NP_004489.1).
Antibody
7L3O1
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Mouse, Rat
ONECUT1
D9Ertd423e; hepatic nuclear factor 6; hepatocyte nuclear factor 6; hepatocyte nuclear factor 6 alpha; hepatocyte nuclear factor 6, alpha; Hfh12; HNF-3/forkhead homolog 12; hnf6; HNF-6; Hnf6a; HNF-6a; Hnf6b; Oc1; OC-1; One cut domain family member 1; one cut domain, family member 1; one cut domain, family member 1, like; one cut homeobox 1; Onecut1; onecut-1 isoform a; onecut-1 isoform b; onecut1 protein; onecut1l; transcription factor; hepatocyte nuclear factor 6; hnf-6; zgc:65904; zgc:77450
ONECUT1
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.