missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HNF6 Recombinant Rabbit Monoclonal Antibody (7L3O1)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558123
This item is not returnable.
View return policy
Description
Immunogen sequence: PYHKDVAGMG QSLSPLSSSG LGSIHNSQQG LPHYAHPGAA MPTDKMLTPN GFEAHHPAML GRHGEQHLTP TSAGMVPING LPPHHPHAHL NAQGHGQLLG TAREPNPSVT GAQVSNGSNS GQMEEI.
Hepatocyte nuclear factor 6 is a CUT homeobox family protein which is a transcription factor and contains a CUT DNA-binding domain and a homeobox DNA-binding domain. It is an upstream regulator of several genes involved in the pathogenesis of maturity-onset diabetes of the young. HNF6 is a ligand of the FOXM1 transcription factor network. HNF6, along with HNF4, controls gene expression in the liver. The expression levels of HNF6 and HNF4 rise steadily during development and that up- or down regulation of HNF6 and HNF4 expression generate premature or delayed expression of the g6pc gene.
Specifications
| HNF6 | |
| Recombinant Monoclonal | |
| 2.29 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| O08755, P70512 | |
| ONECUT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 167-292 of human HNF-6/ONECUT1 (NP_004489.1). | |
| 20 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 7L3O1 | |
| Unconjugated | |
| ONECUT1 | |
| D9Ertd423e; hepatic nuclear factor 6; hepatocyte nuclear factor 6; hepatocyte nuclear factor 6 alpha; hepatocyte nuclear factor 6, alpha; Hfh12; HNF-3/forkhead homolog 12; hnf6; HNF-6; Hnf6a; HNF-6a; Hnf6b; Oc1; OC-1; One cut domain family member 1; one cut domain, family member 1; one cut domain, family member 1, like; one cut homeobox 1; Onecut1; onecut-1 isoform a; onecut-1 isoform b; onecut1 protein; onecut1l; transcription factor; hepatocyte nuclear factor 6; hnf-6; zgc:65904; zgc:77450 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 15379, 25231 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction