missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-6/ONECUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£167.00 - £310.00
Specifications
| Antigen | HNF-6/ONECUT1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18171302
|
Novus Biologicals
NBP2-27079SS |
0.025 mg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18114922
|
Novus Biologicals
NBP2-27079 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HNF-6/ONECUT1 Polyclonal specifically detects HNF-6/ONECUT1 in Human, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HNF-6/ONECUT1 | |
| Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Canine, Primate | |
| Q9UBC0 | |
| 3175 | |
| A portion of amino acids 420-465 of human HNF6 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1 | |
| ONECUT1 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, dog and rat, 94% homologous in cow, 88% homologous in mouse and opossum, and 82% homologous in zebrafish. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title