missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-6/ONECUT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27079SS
This item is not returnable.
View return policy
Description
HNF-6/ONECUT1 Polyclonal specifically detects HNF-6/ONECUT1 in Human, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HNF-6/ONECUT1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3175 | |
| Human, Canine, Primate | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 3-5ug/ml∼, Immunohistochemistry-Paraffin 5ug/ml | |
| Q9UBC0 | |
| ONECUT1 | |
| A portion of amino acids 420-465 of human HNF6 was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey, dog and rat, 94% homologous in cow, 88% homologous in mouse and opossum, and 82% homologous in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction