missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HN1 Recombinant Rabbit Monoclonal Antibody (23GB6065), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | HN1 |
|---|---|
| Clone | 23GB6065 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30088390
|
Invitrogen™
MA552978 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088611
|
Invitrogen™
MA552979 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HN1 Recombinant Monoclonal antibody specifically detects HN1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| HN1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P97825, Q6AXU6, Q9UK76 | |
| 15374, 287828, 51155 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6065 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Androgen-regulated protein 2; ARM2; hematological and neurological expressed 1; hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; HN1; HN1A; Jpt1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed | |
| A synthesized peptide derived from human HN1 (100-154AA). | |
| Primary | |
| -20°C | |
| JPT1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title