missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HN1 Recombinant Rabbit Monoclonal Antibody (23GB6065), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552979
This item is not returnable.
View return policy
Description
HN1 (hematopoietic- and neurologic-expressed sequence 1) is a highly conserved protein that is expressed in developing and regenerating tissues. Overexpression of HN1 has been reported in various tumors like glioma and breast cancer. Hn1 has also been reported as a marker for human ovarian carcinoma and can distinguish epithelial ovarian carcinoma cells from normal ovarian surface epithelial cells. This collective information suggests that HN1 is involved in processes associated with cell proliferation, repair and/or growth.
Specifications
| HN1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| JPT1 | |
| Androgen-regulated protein 2; ARM2; hematological and neurological expressed 1; hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; HN1; HN1A; Jpt1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 15374, 287828, 51155 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6065 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97825, Q6AXU6, Q9UK76 | |
| JPT1 | |
| A synthesized peptide derived from human HN1 (100-154AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering