missing translation for 'onlineSavingsMsg'
Learn More

HMSPP, TRC

Product Code. 30407483
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407483 10 mg 10mg
30401755 25 mg -
30406196 2.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407483 Supplier Toronto Research Chemicals Supplier No. TRCH11502010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 572912-13-1
Molecular Formula C11 H14 O6 S
Molecular Weight (g/mol) 274.2903
Synonym Benzeneacetic acid, 2-hydroxy-α,α-dimethyl-5-[(methylsulfonyl)oxy], HMSPP
IUPAC Name 2-(2-hydroxy-5-methylsulfonyloxyphenyl)-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)c1cc(OS(=O)(=O)C)ccc1O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H14O6S/c1-11(2,10(13)14)8-6-7(4-5-9(8)12)17-18(3,15)16/h4-6,12H,1-3H3,(H,13,14)
Formula Weight 274.0511
Percent Purity >95%
Chemical Name or Material Ethofumesate metabolite NC 20645
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.