CAS RN 572912-13-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
HMSPP, TRC
CAS: 572912-13-1 Molecular Formula: C11 H14 O6 S Molecular Weight (g/mol): 274.2903 Synonym: Benzeneacetic acid, 2-hydroxy-α,α-dimethyl-5-[(methylsulfonyl)oxy],HMSPP IUPAC Name: 2-(2-hydroxy-5-methylsulfonyloxyphenyl)-2-methylpropanoic acid SMILES: CC(C)(C(=O)O)c1cc(OS(=O)(=O)C)ccc1O