missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGXB4 Rabbit anti-Human, Proteintech
Rabbit PolyClonal Antibody
Marca: Proteintech 24821-1-AP-20UL
This item is not returnable.
View return policy
Description
High mobility group (HMG) proteins are nonhistone chromosomal proteins.Specifications
| HMGXB4 | |
| Polyclonal | |
| Unconjugated | |
| HMGXB4 | |
| HMG box containing protein 4, HMG box domain containing 4, HMG2L1, HMGBCG, HMGXB4, Protein HMGBCG, THC211630 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10042 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UGU5 | |
| HMGXB4 | |
| HMGXB4 Fusion Protein Ag20424 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction