missing translation for 'onlineSavingsMsg'
Learn More

HMGXB4 Rabbit anti-Human, Proteintech

Rabbit PolyClonal Antibody

£135.00 - £349.00

Specifications

Antigen HMGXB4
Concentration 0.37 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16883315
View Documents
Proteintech
24821-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16873315
View Documents
Proteintech
24821-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

High mobility group (HMG) proteins are nonhistone chromosomal proteins.
Specifications

Specifications

HMGXB4
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human
Q9UGU5
10042
HMGXB4 Fusion Protein Ag20424
Primary
-20°C
HMGXB4
0.37 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
HMG box containing protein 4, HMG box domain containing 4, HMG2L1, HMGBCG, HMGXB4, Protein HMGBCG, THC211630
HMGXB4
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.