missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3/HMG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | HMGB3/HMG4 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18140292
|
Novus Biologicals
NBP2-24512SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18166105
|
Novus Biologicals
NBP2-24512 |
0.1 mg |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGB3/HMG4 Polyclonal specifically detects HMGB3/HMG4 in Human, Mouse, Bovine, Canine, Chicken, Equine, Primate samples. It is validated for Western Blot.Specifications
| HMGB3/HMG4 | |
| Western Blot 0.5-1.0 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research, DNA Repair | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein | |
| HMGB3 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c). |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Bovine, Canine, Chicken, Equine, Primate | |
| O15347 | |
| 3149 | |
| A portion of amino acids 150-200 of human HMGB3 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title