missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB3/HMG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24512
This item is not returnable.
View return policy
Description
HMGB3/HMG4 Polyclonal specifically detects HMGB3/HMG4 in Human, Mouse, Bovine, Canine, Chicken, Equine, Primate samples. It is validated for Western Blot.
Specifications
| HMGB3/HMG4 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| High mobility group protein 2a, High mobility group protein 4, high mobility group protein B3, high-mobility group (nonhistone chromosomal) protein 4, high-mobility group box 3, HMG-2a, HMG2AHMG-4, HMG4MGC90319, non-histone chromosomal protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_c). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5-1.0 ug/ml | |
| O15347 | |
| HMGB3 | |
| A portion of amino acids 150-200 of human HMGB3 was used as the immunogen. | |
| 0.1 mg | |
| Chromatin Research, DNA Repair | |
| 3149 | |
| Human, Mouse, Bovine, Canine, Chicken, Equine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction