missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGB2 Recombinant Rabbit Monoclonal Antibody (23GB4490), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | HMGB2 |
|---|---|
| Clone | 23GB4490 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087630
![]() |
Invitrogen™
MA553754 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087986
|
Invitrogen™
MA553755 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGB2 Recombinant Monoclonal antibody specifically detects HMGB2 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| HMGB2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P26583, P30681, P52925 | |
| 29395, 3148, 97165 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4490 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| C80539; high mobility group box 2; high mobility group protein 2; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG 2; HMG B2; Hmg2; HMG-2; HMGB2; HMG-B2; HMGB-2 | |
| A synthesized peptide derived from human HMGB2 (1-51AA). | |
| Primary | |
| -20°C | |
| Hmgb2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title