missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGB2 Recombinant Rabbit Monoclonal Antibody (23GB4490), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553755
This item is not returnable.
View return policy
Description
HMGB2 encodes a member of the non-histone chromosomal high mobility group protein family. The proteins of this family are chromatin-associated and ubiquitously distributed in the nucleus of higher eukaryotic cells. In vitro studies have demonstrated that this protein is able to efficiently bend DNA and form DNA circles. These studies suggest a role in facilitating cooperative interactions between cis-acting proteins by promoting DNA flexibility. This protein was also reported to be involved in the final ligation step in DNA end-joining processes of DNA double-strand breaks repair and V(D)J recombination.
Specifications
| HMGB2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Hmgb2 | |
| C80539; high mobility group box 2; high mobility group protein 2; High mobility group protein B2; high-mobility group (nonhistone chromosomal) protein 2; high-mobility group box 2; HMG 2; HMG B2; Hmg2; HMG-2; HMGB2; HMG-B2; HMGB-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29395, 3148, 97165 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4490 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P26583, P30681, P52925 | |
| Hmgb2 | |
| A synthesized peptide derived from human HMGB2 (1-51AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction