missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £309.00
Specifications
| Antigen | HMGB1/HMG-1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 5-10ug/ml∼, Simple Western 1:100, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18066983
|
Novus Biologicals
NBP2-27117SS |
0.025 mg |
£159.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767853
|
Novus Biologicals
NBP2-27117 |
0.1 mg |
£309.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HMGB1/HMG-1 | |
| Western Blot 5-10ug/ml∼, Simple Western 1:100, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| P09429 | |
| 3146 | |
| A portion of amino acids 100-166 of human HMGB1 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| HMGB1 | |
| IgG | |
| Protein A purified | |
| The HMGB1 amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chicken, cow and monkey. This antibody recognizes both B box domain (amino acids 88-162) and full length HMGB1 protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title