missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27117SS
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HMGB1/HMG-1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The HMGB1 amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chicken, cow and monkey. This antibody recognizes both B box domain (amino acids 88-162) and full length HMGB1 protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 5-10ug/ml∼, Simple Western 1:100, Flow Cytometry 0.5ug/10^6cells, Immunohistochemistry-Paraffin 5ug/ml | |
| P09429 | |
| HMGB1 | |
| A portion of amino acids 100-166 of human HMGB1 was used as the immunogen. | |
| 0.025 mg | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| 3146 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction