missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
£215.00 - £493.00
Specifications
| Antigen | HMGB1/HMG-1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1.0 ug/ml, Simple Western 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 0.05 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Flow (Intracellular) |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18796663
|
Novus Biologicals
NB100-2322SS |
0.025 mL |
£215.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18768703
|
Novus Biologicals
NB100-2322 |
£493.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.Specifications
| HMGB1/HMG-1 | |
| Western Blot 0.5-1.0 ug/ml, Simple Western 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 0.05 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Flow (Intracellular) | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| Tris-Glycine with 0.02% Sodium Azide | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| HMGB1 | |
| IgG | |
| Affinity Purified | |
| Human, mouse, rat, dog, bovine, sheep. Predicted to react with porcine, equine and rabbit based on 100% sequence homology. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep | |
| P09429 | |
| 3146 | |
| A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 29 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title