missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 22 publications
Brand: Novus Biologicals NB100-2322SS
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Polyclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.
Specifications
| HMGB1/HMG-1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Glycine with 0.02% Sodium Azide | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| Rabbit | |
| 29 kDa | |
| 0.025 mL | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| 3146 | |
| Human, Mouse, Rat, Bovine, Canine, Rabbit, Sheep | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-1.0 ug/ml, Simple Western 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:100-1:250, Immunocytochemistry/Immunofluorescence 0.05 ug/ml, Immunohistochemistry-Paraffin 1:100-1:250, Flow (Intracellular) | |
| P09429 | |
| HMGB1 | |
| A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse, rat, dog, bovine, sheep. Predicted to react with porcine, equine and rabbit based on 100% sequence homology. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction