missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14582-1-AP-150UL
This item is not returnable.
View return policy
Description
Mutations of the human BRCA2 gene confer susceptibility to breast cancer. A structural DNA binding component has been identified and is designated as BRCA2-Associated Factor 35 (BRAF35). There is a close association of the BRAF35/BRCA2 complex with condensed chromatin coincident with histone H3 phosphorylation. Experimental evidence suggests a role for the BRCA2/BRAF35 complex in the modulation of cell cycle progression.Specifications
| HMG20B | |
| Polyclonal | |
| Unconjugated | |
| HMG20B | |
| BRAF25, BRAF35, BRCA2 associated factor 35, high mobility group 20B, HMG box containing protein 20B, HMG20B, HMGX2, HMGXB2, PP7706, pp8857, SMARCE1 related protein, SMARCE1r, SOXL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10362, 15353, 362825 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9P0W2, Q9Z104 | |
| Hmg20b | |
| HMG20B Fusion Protein Ag6103 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido