missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HMG20B |
|---|---|
| Concentration | 0.23 mg/mL |
| Applications | Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803294
|
Proteintech
14582-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893284
|
Proteintech
14582-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mutations of the human BRCA2 gene confer susceptibility to breast cancer. A structural DNA binding component has been identified and is designated as BRCA2-Associated Factor 35 (BRAF35). There is a close association of the BRAF35/BRCA2 complex with condensed chromatin coincident with histone H3 phosphorylation. Experimental evidence suggests a role for the BRCA2/BRAF35 complex in the modulation of cell cycle progression.Specifications
| HMG20B | |
| Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| Q9P0W2, Q9Z104 | |
| 10362, 15353, 362825 | |
| HMG20B Fusion Protein Ag6103 | |
| Primary | |
| -20°C | |
| HMG20B |
| 0.23 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| BRAF25, BRAF35, BRCA2 associated factor 35, high mobility group 20B, HMG box containing protein 20B, HMG20B, HMGX2, HMGXB2, PP7706, pp8857, SMARCE1 related protein, SMARCE1r, SOXL | |
| Hmg20b | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title