missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Mouse anti-Human, Mouse, Clone: 1B10F12, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | HMG20A |
|---|---|
| Clone | 1B10F12 |
| Concentration | 1 mg/mL |
| Applications | Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888775
|
Proteintech
66676-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878775
|
Proteintech
66676-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.Specifications
| HMG20A | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| FLJ10739, high mobility group 20A, HMG box containing protein 20A, HMG20A, HMGX1, HMGXB1 | |
| Hmg20a | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1B10F12 | |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q9DC33, Q9NP66 | |
| 10363, 66867 | |
| HMG20A Fusion Protein Ag2718 | |
| Primary | |
| -20°C | |
| HMG20A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title