missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Mouse anti-Human, Mouse, Clone: 1B10F12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66676-1-IG-20UL
This item is not returnable.
View return policy
Description
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.Specifications
| HMG20A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9DC33, Q9NP66 | |
| Hmg20a | |
| HMG20A Fusion Protein Ag2718 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 1B10F12 | |
| Unconjugated | |
| HMG20A | |
| FLJ10739, high mobility group 20A, HMG box containing protein 20A, HMG20A, HMGX1, HMGXB1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 10363, 66867 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction