missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HM 650V Vibrating Blade Microtome Specimen Stage F (Flat)for medium to large specimens or tissue (from 10mm to 45mm)
Use these Epredia™ HM 650V Vibrating Blade Microtome Accessories with the Epredia™ HM650V Vibrating Blade Microtome.
£57.00 - £185.00
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12013029
|
Epredia™
152510 |
Adapter I |
£185.00
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12023029
|
Epredia™
152520 |
Adapter S |
£99.25
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12033029
|
Epredia™
152530 |
Adapter R |
£77.00
Each |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12067229
|
Epredia™
715810 |
Specimen Stage F (Flat) |
£57.00
Each |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Standard buffer tray:- Hold stages in place using a magnet
- Fill tray with all water-based buffers and solutions
- Use pre-cooled in the standard buffer tray for cooling tissue
- Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
- Magnification: 2.5x
- Ability to move, swivel, and displace for optimal viewing position
- Adapter and illumination included
- Eyepieces: 10x
- Supplementary lens: 0.63x
- Zoom magnifying range: 4.1 to 31.5x
- Microscope adapter for angled view of specimen
- Horizontal view and vertical drive and swivel mechanism
- 3.5mm fiber bundle
- 50mm long swan-neck light guide
- 750mm long flexible light guide
- High light intensity
- Adapter for the KL200 (760317) included