missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ HM 650V Vibrating Blade Microtome Specimen Stage F (Flat)for medium to large specimens or tissue (from 10mm to 45mm)
Use these Epredia™ HM 650V Vibrating Blade Microtome Accessories with the Epredia™ HM650V Vibrating Blade Microtome.
Brand: Epredia™ 152520
Code : 50
Additional Details : Weight : 2.00000kg
Description
Standard buffer tray:- Hold stages in place using a magnet
- Fill tray with all water-based buffers and solutions
- Use pre-cooled in the standard buffer tray for cooling tissue
- Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
- Magnification: 2.5x
- Ability to move, swivel, and displace for optimal viewing position
- Adapter and illumination included
- Eyepieces: 10x
- Supplementary lens: 0.63x
- Zoom magnifying range: 4.1 to 31.5x
- Microscope adapter for angled view of specimen
- Horizontal view and vertical drive and swivel mechanism
- 3.5mm fiber bundle
- 50mm long swan-neck light guide
- 750mm long flexible light guide
- High light intensity
- Adapter for the KL200 (760317) included
Specifications
Adapter S | |
Adapter S | |
Sapphire and Zirconium Blades |