Learn More
HLA-E Monoclonal Antibody (3D12HLA-E), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£132.00 - £210.00
Specifications
Antigen | HLA-E |
---|---|
Clone | 3D12HLA-E |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15588276
|
Affymetrix eBioscience
17-9953-42 |
100 Tests |
£210.00
100 tests |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578276
|
Affymetrix eBioscience
17-9953-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal antibody 3D12 (HLA-E) recognizes human HLA-E, a member of the Human Leukocyte Antigen family. HLA-E is a MHC Class IB member, which has a specialized role in NK cell recognition. Binding of a restricted set of peptides derived from other Class I leader sequences to the HLA-E and B2M (beta2 microglobulin) complex leads to interaction with NKG2A on NK cells. This binding protects the HLA-G-expressing cell from NK cell killing. HLA-E expression is ubiquitous. Applications Reported: This 3D12HLA-E antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3D12HLA-E antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
HLA-E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-E binds a restricted subset of peptides derived from the leader peptides of other class I molecules. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
HLA-E | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
DAQB-90C11.16, FLJ26655, HLAA | |
HLA-E | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
HLA-E |
3D12HLA-E | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
P13747 | |
3133 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.