missing translation for 'onlineSavingsMsg'
Learn More

HLA-E Monoclonal Antibody (3D12HLA-E), APC, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-9953-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15588276

  • £210.00 / 100 tests
Estimated Shipment: 13-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody 3D12 (HLA-E) recognizes human HLA-E, a member of the Human Leukocyte Antigen family. HLA-E is a MHC Class IB member, which has a specialized role in NK cell recognition. Binding of a restricted set of peptides derived from other Class I leader sequences to the HLA-E and B2M (beta2 microglobulin) complex leads to interaction with NKG2A on NK cells. This binding protects the HLA-G-expressing cell from NK cell killing. HLA-E expression is ubiquitous. Applications Reported: This 3D12HLA-E antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3D12HLA-E antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.5 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

HLA-E belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. HLA-E binds a restricted subset of peptides derived from the leader peptides of other class I molecules. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications

Specifications

HLA-E
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
P13747
HLA-E
Affinity chromatography
RUO
3133
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
3D12HLA-E
APC
HLA-E
A29 protein; antigen presenting molecule; Aw-33; Aw-74; DAMA-277I14.1; DAQB-90C11.16; DKFZp686P19218; EA1.2; EA2.1; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, alpha chain E; HLA class I histocompatibility antigen, E alpha chain; HLA locus; HLA-6.2; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; HLAE; HLA-E; Human leukocyte antigen A; leukocyte antigen class I-A; lymphocyte antigen; major histocompatibility complex, class I, A; major histocompatibility complex, class I, E; MCH class I antigen; MHC; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; M
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.