missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Mouse, PE, Clone: L243, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio QAB86-PE-25Tests

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15942578

  • £89.95 / 25 tests
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HLA-DR
Monoclonal
5μL/Test
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
HLA-DRA
Protein A/G Purified
RUO
3122
IgG2a κ
Flow Cytometry
L243
PE
NP_061984
Mouse
25 Tests
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers