missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Mouse, PE, Clone: L243, enQuireBio™
Mouse Monoclonal Antibody
£89.95 - £920.00
Specifications
Antigen | HLA-DR |
---|---|
Clone | L243 |
Concentration | 5μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942578
|
EnquireBio
QAB86-PE-25TESTS |
25 Tests |
£89.95
25 tests |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15932578
|
EnquireBio
QAB86-PE-100TESTS |
100 Tests |
£210.00
100 tests |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15952578
|
EnquireBio
QAB86-PE-500TESTS |
500 Tests |
£920.00
500 tests |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HLA-DR | |
5μL/Test | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
3122 | |
IgG2a κ | |
Protein A/G Purified |
L243 | |
Flow Cytometry | |
PE | |
RUO | |
NP_061984 | |
HLA-DRA | |
Primary | |
Store at 2-4°C. |