missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma,Dog, BB700, Clone: G46-6, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566481
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human,Baboon,Cynomolgus,Rhesus Ma,Dog samples. It is validated for Flow Cytometry.
Specifications
| HLA-DR | |
| Monoclonal | |
| Brilliant Blue 700 (BB700) | |
| 25 Tests | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
| Flow Cytometry | |
| G46-6 | |
| Mouse | |
| RUO | |
| Human, Baboon, Cynomolgus, Rhesus Macaque, Dog | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction