missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Mouse anti-Human,Baboon,Cynomolgus,Rhesus Ma,Dog, BB700, Clone: G46-6, BD Horizon™
Mouse Monoclonal Antibody
£187.38 - £389.82
Specifications
| Antigen | HLA-DR |
|---|---|
| Clone | G46-6 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 700 (BB700) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15855089
|
BD Biosciences
566481 |
25 Tests |
£196.00 £187.38 / Each Save £8.62 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15845089
|
BD Biosciences
566480 |
100 Tests |
£402.00 £389.82 / Each Save £12.18 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human,Baboon,Cynomolgus,Rhesus Ma,Dog samples. It is validated for Flow Cytometry.Specifications
| HLA-DR | |
| Flow Cytometry | |
| Brilliant Blue 700 (BB700) | |
| Mouse | |
| Human, Baboon, Cynomolgus, Rhesus Macaque, Dog | |
| Primary |
| G46-6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| IgG2a κ | |
| Refrigerate (2 to 8°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title